Bismuth 2-ethylhexanoate
Bismuth 2-ethylhexanoate
Bismuth 2-ethylhexanoate
Thermo Scientific Chemicals

Bismuth 2-ethylhexanoate

CAS: 67874-71-9 | C24H48BiO6 | 641.622 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 036710.14
also known as 036710-14
Price (EUR)
151,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
151,00
Each
Chemical Identifiers
CAS67874-71-9
IUPAC Namebismuth(3+) tris(2-ethylhexanoate)
Molecular FormulaC24H45BiO6
InChI KeyNUMHJBONQMZPBW-UHFFFAOYNA-K
SMILES[Bi+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
Assay from Supplier's CofA≥60%
FormViscous liquid
Bismuth 2-ethylhexanoate is used as an industrial lubricant and dessicant. It is also used for synthesizing of urethane foam, urethane coating and other urethane products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only