Iridium, 0.5% on activated carbon powder, reduced, nominally 50% water wet
Iridium, 0.5% on activated carbon powder, reduced, nominally 50% water wet
Iridium, 0.5% on activated carbon powder, reduced, nominally 50% water wet
Thermo Scientific Chemicals

Iridium, 0.5% on activated carbon powder, reduced, nominally 50% water wet

CAS: 7439-88-5 | Ir | 192.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 038327.06
also known as 038327-06
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS7439-88-5
ChEBICHEBI:49666
Chemical Name or MaterialIridium
IUPAC Nameiridium
InChI KeyGKOZUEZYRPOHIO-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only