Cadmium sulfide, 99.999% (metals basis)
Cadmium sulfide, 99.999% (metals basis)
Cadmium sulfide, 99.999% (metals basis)
Thermo Scientific Chemicals

Cadmium sulfide, 99.999% (metals basis)

CAS: 1306-23-6 | CdS | 144.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 038342.30
also known as 038342-30
Price (EUR)
719,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
719,00
Each
Chemical Identifiers
CAS1306-23-6
IUPAC Namecadmium(2+) sulfanediide
Molecular FormulaCdS
InChI KeyFRLJSGOEGLARCA-UHFFFAOYSA-N
SMILES[S--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
The main application of cadmium sulfide is as a pigment. Cadmium sulfide is also used in the production of solar cells. Used as light dependent resistors/photo resistors for light sensors. Used in MOCVD, sputtering and electrochemical deposition. In screen printing, it is used as a slurry containing dispersed cadmium sulfide. It is also used in dandruff shampoos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only