Barium fluoride crystal optic disc, 32mm x 3mm, polished both sides
Thermo Scientific Chemicals

Barium fluoride crystal optic disc, 32mm x 3mm, polished both sides

 
Have Questions?
Catalog NumberQuantity
038436.KT1 Ea.
Catalog number 038436.KT
Price (EUR)
546,00
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (EUR)
546,00
Each
Specifications
Chemical Name or MaterialBarium fluoride crystal optic disc
EINECS Number232-108-0
Health Hazard 1H302-H332
Health Hazard 3P261-P264-P304+P340-P301+P312-P312-P501a
Name Note32mm x 3mm, Polished both sides
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only