Vanadium wire, 2.0mm (0.08 in.) dia., 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium wire, 2.0mm (0.08 in.) dia., 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium wire, 2.0mm (0.08 in.) dia., 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium wire, 2.0mm (0.08 in.) dia., 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
038516.G1
also known as 038516-G1
1 m
Catalog number 038516.G1
also known as 038516-G1
Price (EUR)
-
Quantity:
1 m
Request bulk or custom format
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White silver
FormWire, 2.0mm (0.08in) diameter.
Assay from Supplier's CofA≥99.5% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only