Magnesium phthalocyanine, tech. 90%
Magnesium phthalocyanine, tech. 90%
Magnesium phthalocyanine, tech. 90%
Thermo Scientific Chemicals

Magnesium phthalocyanine, tech. 90%

CAS: 1661-03-6 | C32H18MgN8O | 554.856 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 038601.03
also known as 038601-03
Price (EUR)
55,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
55,40
Each
Specifications
CAS1661-03-6
Chemical Name or MaterialMagnesium phthalocyanine
EINECS Number216-766-6
Formula Weight536.84
Health Hazard 1H250-H314-H335
View more
Magnesium phthalocyanine is used in preparing membrane sensors for cyanide ions. Also used in preparation of membrane sensors for cyanide ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only