Barium fluoride, ultra dry, 99.998% (metals basis)
Barium fluoride, ultra dry, 99.998% (metals basis)
Barium fluoride, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals

Barium fluoride, ultra dry, 99.998% (metals basis)

CAS: 7787-32-8 | BaF2 | 175.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 038610.09
Price (EUR)
104,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
104,00
Each
Chemical Identifiers
CAS7787-32-8
IUPAC Namebarium(2+) difluoride
Molecular FormulaBaF2
InChI KeyOYLGJCQECKOTOL-UHFFFAOYSA-L
SMILES[F-].[F-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Residual water< 100 ppm
Total Metal Impurities0.002% max.
Barium Fluoride is used in ceramic flux, carbon brushes for electrical equipment, glass making, in the manufacture of other fluorides, crystals for spectroscopy, electronics and dry-film lubricants. Barium Fluoride is used in spectroscopic components. Barium Fluoride is often suitable for applications in the passive IR band (8 to 14 ?m) and is often used as a viewport window for thermography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only