Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable
Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable
Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable
Thermo Scientific Chemicals

Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable

CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 038711.K2
also known as 038711-K2
Price (EUR)
50,80
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
50,80
Each
Specifications
CAS1336-21-6
ChEBICHEBI:18219
Chemical Name or MaterialBuffer solution
ColorGreen
IUPAC Nameamine hydrate
View more
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only