Titanium, plasma standard solution, Specpure™ Ti 1000μg/mL
Thermo Scientific Chemicals

Titanium, plasma standard solution, Specpure™ Ti 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 038720.AD
also known as 038720-AD
Price (EUR)
-
Quantity:
50 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTitanium plasma standard solution
MDL NumberMFCD00011264
Name NoteTi 1000μg/mL
Physical FormLiquid
Quantity50 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only