Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
Thermo Scientific Chemicals

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)

CAS: 31277-98-2 | C52H48P4Pd | 903.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
1 g
Catalog number 039233.02
also known as 039233-02
Price (EUR)
97,50
Each
Quantity:
0.5 g
Request bulk or custom format
Price (EUR)
97,50
Each
Chemical Identifiers
CAS31277-98-2
IUPAC Namebis([2-(diphenylphosphanyl)ethyl]diphenylphosphane) palladium
Molecular FormulaC52H48P4Pd
InChI KeyFAFGMAGIYHHRKN-UHFFFAOYSA-N
SMILES[Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only