Dichlorobis(ethylenediamine)platinum(II), Pt 50%
Dichlorobis(ethylenediamine)platinum(II), Pt 50%
Dichlorobis(ethylenediamine)platinum(II), Pt 50%
Dichlorobis(ethylenediamine)platinum(II), Pt 50%
Dichlorobis(ethylenediamine)platinum(II), Pt 50%
Dichlorobis(ethylenediamine)platinum(II), Pt 50%
Dichlorobis(ethylenediamine)platinum(II), Pt 50%
Thermo Scientific Chemicals

Dichlorobis(ethylenediamine)platinum(II), Pt 50%

CAS: 21430-85-3 | C4H16Cl2N4Pt | 386.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 039280.03
also known as 039280-03
Price (EUR)
261,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
261,00
Each
Specifications
CAS21430-85-3
Chemical Name or MaterialDichlorobis(ethylenediamine)platinum(II)
ColorWhite
Concentration or Composition (by Analyte or Components)Pt 50%
Formula Weight386.18
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only