Tris(isopropylcyclopentadienyl)lanthanum(III), 99.9% (REO)
Tris(isopropylcyclopentadienyl)lanthanum(III), 99.9% (REO)
Tris(isopropylcyclopentadienyl)lanthanum(III), 99.9% (REO)
Thermo Scientific Chemicals

Tris(isopropylcyclopentadienyl)lanthanum(III), 99.9% (REO)

CAS: 68959-87-5 | C24H33La | 460.43 g/mol
Have Questions?
Catalog NumberQuantity
039323.03
also known as 039323-03
1 g
Catalog number 039323.03
also known as 039323-03
Price (EUR)
107,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
107,00
Each
Specifications
Assay Percent Notes(REO)
CAS68959-87-5
Chemical Name or MaterialTris(isopropylcyclopentadienyl)lanthanum(III)
ColorColorless to Yellow
Formula Weight460.43
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only