Lithium 2-ethylhexanoate
Lithium 2-ethylhexanoate
Lithium 2-ethylhexanoate
Thermo Scientific Chemicals

Lithium 2-ethylhexanoate

CAS: 15590-62-2 | C8H15LiO2 | 150.146 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 039330.09
also known as 039330-09
Price (EUR)
64,60
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
64,60
Each
Specifications
CAS15590-62-2
Chemical Name or MaterialLithium 2-ethylhexanoate
EINECS Number239-657-5
Formula Weight150.15
Health Hazard 1H361d
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only