Praseodymium(III) chloride, ultra dry, 99.99% (REO)
Praseodymium(III) chloride, ultra dry, 99.99% (REO)
Praseodymium(III) chloride, ultra dry, 99.99% (REO)
Thermo Scientific Chemicals

Praseodymium(III) chloride, ultra dry, 99.99% (REO)

CAS: 10361-79-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 039386.06
also known as 039386-06
Price (EUR)
157,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
157,00
Each
Chemical Identifiers
CAS10361-79-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
FormPowder
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.01% max.
Praseodymium(III) chloride is used in producing metal praseodymium and praseodymium compounds,glass,ceramic and coloring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only