Potassium manganate
Potassium manganate
Potassium manganate
Potassium manganate
Thermo Scientific Chemicals

Potassium manganate

CAS: 10294-64-1 | K2MnO4 | 197.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 039506.09
also known as 039506-09
Price (EUR)
76,33
Online Exclusive
89,80
Save 13,47 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)
76,33
Online Exclusive
89,80
Save 13,47 (15%)
Each
Add to cart
Chemical Identifiers
CAS10294-64-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark purple to dark green to black
Assay from Suppliers CofA≥94%
FormPowder or crystalline powder or crystals
It serves as an oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only