Tin(II) phthalocyanine, Thermo Scientific Chemicals
Tin(II) phthalocyanine, Thermo Scientific Chemicals
Tin(II) phthalocyanine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) phthalocyanine, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
039607.02
also known as 039607-02
0.5 g
Catalog number 039607.02
also known as 039607-02
Price (EUR)
74,90
Each
Quantity:
0.5 g
Request bulk or custom format
Price (EUR)
74,90
Each
Specifications
CAS15304-57-1
Chemical Name or MaterialTin(II) phthalocyanine
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Formula Weight631.43
Health Hazard 1H302+H312+H332-H315-H319-H335
View more
Tin(II) phthalocyanine is used as an organic light-emitting diode (OLED) material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only