Sodium stearate
Sodium stearate
Sodium stearate
Thermo Scientific Chemicals

Sodium stearate

CAS: 822-16-2 | C18H35NaO2 | 306.466 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 039648.A7
also known as 039648-A7
Price (EUR)
223,65
Online exclusive
248,00
Save 24,35 (10%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (EUR)
223,65
Online exclusive
248,00
Save 24,35 (10%)
Each
Chemical Identifiers
CAS822-16-2
IUPAC Namesodium octadecanoate
Molecular FormulaC18H35NaO2
InChI KeyRYYKJJJTJZKILX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
CommentFree Fatty Acid (FFA): ≤3.0%
CommentMelting Point: ≥200°C
Loss on Drying≤3.0%
Appearance (Color)White
Used in adhesives and sealants, laundry and dishwashing products, plastic and rubber products. Used as surface active agents. It is the gelling agent for deodrant sticks. Used as waterproofing additives and ointments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only