Triethyl phosphate, 99+%
Triethyl phosphate, 99+%
Triethyl phosphate, 99+%
Triethyl phosphate, 99+%
Triethyl phosphate, 99+%
Thermo Scientific Chemicals

Triethyl phosphate, 99+%

CAS: 78-40-0 | C6H15O4P | 182.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 040001.22
also known as 040001-22
Price (EUR)
49,22
온라인 행사
57,90
Save 8,68 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
49,22
온라인 행사
57,90
Save 8,68 (15%)
Each
Add to cart
Specifications
CAS78-40-0
ChEBICHEBI:45927
Chemical Name or MaterialTriethyl phosphate
Density1.072 g/mL
EINECS Number201-114-5
View more
Triethyl phosphate is used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only