Potassium hexafluorophosphate, 99% min
Potassium hexafluorophosphate, 99% min
Potassium hexafluorophosphate, 99% min
Thermo Scientific Chemicals

Potassium hexafluorophosphate, 99% min

CAS: 17084-13-8 | F6KP | 184.062 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
500 g
Catalog number 040030.36
also known as 040030-36
Price (EUR)
201,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
201,00
Each
Chemical Identifiers
CAS17084-13-8
IUPAC Namepotassium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6KP
InChI KeyYZDGRYDIGCWVND-UHFFFAOYSA-N
SMILES[K+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAFree fluoride: ≤0.5%
Water Content≤1.0%
FormPowder or crystals or crystalline powder
Assay from Supplier's CofAPF₆ anion: ≥77.6% (theoretical 78.8%)
Appearance (Color)White
Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only