Silver(II) oxide, 99.9% (metals basis), Ag 86.6% min
Silver(II) oxide, 99.9% (metals basis), Ag 86.6% min
Silver(II) oxide, 99.9% (metals basis), Ag 86.6% min
Thermo Scientific Chemicals

Silver(II) oxide, 99.9% (metals basis), Ag 86.6% min

CAS: 1301-96-8 | AgO | 123.867 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 040187.14
also known as 040187-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1301-96-8
IUPAC Nameoxosilver
Molecular FormulaAgO
InChI KeyOTCVAHKKMMUFAY-UHFFFAOYSA-N
SMILESO=[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Transition TemperatureAg 86.6% min
Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only