Copper(I) oxide, 99% (metals basis)
Copper(I) oxide, 99% (metals basis)
Copper(I) oxide, 99% (metals basis)
Thermo Scientific™

Copper(I) oxide, 99% (metals basis)

CAS: 1317-39-1 | Cu2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 040188.14
also known as 040188-14
Price (EUR)
37,20
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
37,20
Each
Specifications
AppearancePurple-red powder
Assay Percent Range99% (Metal basis)
Boiling Point1800°C (decomposition)
CAS1317-39-1
Chemical Name or MaterialCopper(I) oxide
View more
Used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only