Praseodymium rod, 6.35mm (0.25in) dia, 98.5% (metals basis excluding Ta)
Praseodymium rod, 6.35mm (0.25in) dia, 98.5% (metals basis excluding Ta)
Praseodymium rod, 6.35mm (0.25in) dia, 98.5% (metals basis excluding Ta)
Thermo Scientific Chemicals

Praseodymium rod, 6.35mm (0.25in) dia, 98.5% (metals basis excluding Ta)

Form: Approx. 0.22g/mm, Cast rod, diameter is approximate | CAS: 7440-10-0 | Pr | 140.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
50 mm
100 mm
Catalog number 040295.DJ
also known as 040295-DJ
Price (EUR)
435,00
Each
Quantity:
50 mm
Request bulk or custom format
Price (EUR)
435,00
Each
Chemical Identifiers
CAS7440-10-0
IUPAC Namepraseodymium
Molecular FormulaPr
InChI KeyPUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES[Pr]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod
Appearance (Color)Silver
Total Metal Impurities excluding Ta≤1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only