Hydrogen tetranitratoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 38.9-40.1%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrogen tetranitratoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 38.9-40.1%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
040431.06
also known as 040431-06
5 g
Catalog number 040431.06
also known as 040431-06
Price (EUR)
834,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
834,00
Each
Specifications
Assay Percent Notes(metals basis)
Chemical Name or MaterialHydrogen tetranitratoaurate(III) hydrate
Concentration or Composition (by Analyte or Components)Au 38.9 to 40.1%
DOT InformationTransport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
Formula Weight445.99 (Anhydrous)
View more
Used for Isomerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only