Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol
Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol
Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol
Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol
Thermo Scientific Chemicals

Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol

CAS: 876107-33-4 | C12H28CeO8 | 440.464 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 040471.06
also known as 040471-06
Price (EUR)
78,29
キャンペーン価格
92,10
Save 13,81 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
78,29
キャンペーン価格
92,10
Save 13,81 (15%)
Each
Add to cart
Specifications
CAS876107-33-4
Chemical Name or MaterialCerium(IV) 2-methoxyethoxide
ColorYellow
Concentration or Composition (by Analyte or Components)18 to 20% w/w in 2-methoxyethanol
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
View more
Hydrolysis of cerium 2-methoxyethoxide and subsequent supercritical drying or freeze-drying yielded a CeO 2 aerogel or cryogel with quite a large surface area above 300 m 2 /g.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only