Ammonium sulfide, 40-44% w/w aq. soln.
Ammonium sulfide, 40-44% w/w aq. soln.
Ammonium sulfide, 40-44% w/w aq. soln.
Thermo Scientific Chemicals

Ammonium sulfide, 40-44% w/w aq. soln.

CAS: 12135-76-1 | H8N2S | 68.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 040509.AP
also known as 040509-AP
Price (EUR)
323,00
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
323,00
Each
Chemical Identifiers
CAS12135-76-1
IUPAC Namediamine sulfane
Molecular FormulaH8N2S
InChI KeyUXXGWUQNRMPNKH-UHFFFAOYSA-N
SMILESN.N.S
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)40-44%
Ammonium sulfide solution is used occasionally in photographic developing, to apply patina to bronze, and in textile manufacturing. Ammonium sulfide is a very useful reagent in mineral analysis. Used in continuous culture of saccharomyces cerevisiae, useful in a double staining technique for visualization of astrocytes and microglia from brain sections and astroglial cell cultures. It plays an important role to modify the surface of low κ dielectric thin films. It is essential for molecular beam epitaxy regrowth.It is an unstable salt and acts as a selective reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only