N-Ethylmaleimide, 99+%
N-Ethylmaleimide, 99+%
N-Ethylmaleimide, 99+%
Thermo Scientific Chemicals

N-Ethylmaleimide, 99+%

CAS: 128-53-0 | C6H7NO2 | 125.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 040526.03
also known as 040526-03
Price (EUR)
46,80
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
46,80
Each
Chemical Identifiers
CAS128-53-0
IUPAC Name1-ethyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular FormulaC6H7NO2
InChI KeyHDFGOPSGAURCEO-UHFFFAOYSA-N
SMILESCCN1C(=O)C=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder and/or chunks
Assay from Supplier's CofA≥99.0% (GC)
Melting Point43-47?C
N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+ dependent internucleosomal DNA fragmentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only