Polyethyleneimine, branched, M.W. 2,000
Polyethyleneimine, branched, M.W. 2,000
Polyethyleneimine, branched, M.W. 2,000
Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 2,000

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 040528.22
also known as 040528-22
Price (EUR)
157,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
157,00
Each
Chemical Identifiers
CAS9002-98-6
Molecular Formula((C2H5N)zC2H4N)y(C2H5N)x
SMILES*-CCNCCN(-*)CCN-*
MDL NumberMFCD00084427
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Colorless to yellow, clear to slightly turbid
Molecular Weight1600-2400
Polyethyleneimine is used for various applications in products like detergents, adhesives, water treatment agents and cosmetics. It is also used as flocculating agent, enzyme carrier, wet-strength agent in the paper-making process, chelating agent form complexes with metal ions such as zinc and zirconium. It acts as scavenger for aldehydes and oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only