Molybdenum wire, gold plated, 0.025mm (0.001in) dia, 99.95% (metals basis)
Molybdenum wire, gold plated, 0.025mm (0.001in) dia, 99.95% (metals basis)
Molybdenum wire, gold plated, 0.025mm (0.001in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum wire, gold plated, 0.025mm (0.001in) dia, 99.95% (metals basis)

Microelectronics, high temperature | CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
10 m
Catalog number 040671.G5
also known as 040671-G5
Price (EUR)
347,00
Each
Quantity:
5 m
Request bulk or custom format
Price (EUR)
347,00
Each
Specifications
Assay Percent Notes(metals basis)
CAS7439-98-7
ChEBICHEBI:28685
Chemical Name or MaterialMolybdenum wire, gold plated
EINECS Number231-107-2
View more
Microelectronics, high temperature

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only