Carbon sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.999% (metals basis)
Thermo Scientific Chemicals

Carbon sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.999% (metals basis)

 
Have Questions?
Catalog NumberQuantity
040832.KS1 Ea.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040832.KS
Price (EUR)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Chemical Name or MaterialCarbon sputtering target
EINECS Number231-955-3
MDL NumberMFCD00133992
Name Note50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
View more
Used as Magnetron sputtering source. The carbon sputtering target was replaced after the de- positions of 20-30 films with several thousand A in thick- ness in mass-separated negative-ion-beam deposition system. Carbon films can also be deposited by magnetron sputtering in which a magnetically confined plasma torus (race track) acts as a distributed source of ions that impact a carbon sputtering target over a significant part of its area. C -ions were produced from a highly pure carbon-sputtering target by cesium ion sputtering in a neutral and ionized alkaline bombardment-type heavy negative ion source. CN negative ions of 0.88 mA were safely obtained by using a carbon sputtering target and nitrogen gas instead of cyanogen for ionization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only