Manganese sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Manganese sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Manganese sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Manganese sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Catalog NumberQuantity
040864.KS1 Ea.
Catalog number 040864.KS
Price (EUR)
1.322,00
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (EUR)
1.322,00
Each
Chemical Identifiers
CAS7439-96-5
IUPAC Namemanganese
Molecular FormulaMn
InChI KeyPWHULOQIROXLJO-UHFFFAOYSA-N
SMILES[Mn]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.95% (metals basis)
FormSputtering target, 76.2mm (3.0in) diameter x 3.18mm (0.125in) thick.
Appearance (Color)Gray
Manganese sputtering target is widely used in coating processing industries like magnetic data storage application, electronic and semiconductor application. It is used also in chemical vapor deposition (CVD), physical vapor deposition (PVD) display and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only