Silver gauze, 50 mesh woven from 0.0764mm (0.003in) dia wire
Silver gauze, 50 mesh woven from 0.0764mm (0.003in) dia wire
Silver gauze, 50 mesh woven from 0.0764mm (0.003in) dia wire
Thermo Scientific Chemicals

Silver gauze, 50 mesh woven from 0.0764mm (0.003in) dia wire

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 040935.FL
also known as 040935-FL
Price (EUR)
-
Quantity:
75 x 75 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment50 mesh woven from 0.0764mm diameter wire
Appearance (Color)Silver
FormGauze
Silver gauze is used for filtration. It is also used in electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only