Stainless Steel gauze, 20 mesh woven from 0.102mm (0.004in) dia wire, Type 316
Thermo Scientific Chemicals

Stainless Steel gauze, 20 mesh woven from 0.102mm (0.004in) dia wire, Type 316

Open area: 84.6%; Width of opening: 0.046 in. | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 040943.FL
also known as 040943-FL
Price (EUR)
41,90
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (EUR)
41,90
Each
Specifications
AppearanceGauze
Chemical Name or MaterialStainless Steel
Health Hazard 1H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
MDL NumberMFCD00148499
Mesh Size20 Mesh
View more
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only