Silver, solder alloy, 1.6mm (0.06in) dia
Thermo Scientific Chemicals

Silver, solder alloy, 1.6mm (0.06in) dia

For use on copper, stainless, and most other metals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
Catalog number 041032.36
also known as 041032-36
Price (EUR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeAg:Sn; 3.5:96.5 wt%
Chemical Name or MaterialSilver, solder Alloy
MDL NumberMFCD00801108
Name Note1.6mm (0.06 in.) dia.
Recommended StorageAmbient temperatures
View more
Silver, solder alloy is used on copper, stainless, and most other metals. It is used for electronic assemblies, die attachments, thick films. Soldering of jewellery due to the alloys ability to maintain high levels of brightness.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only