Silicon(II) oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.9% (metals basis)
Silicon(II) oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.9% (metals basis)
Silicon(II) oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Silicon(II) oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.9% (metals basis)

CAS: 10097-28-6 | OSi | 44.084 g/mol
Have Questions?
Catalog NumberQuantity
041093.KS1 Ea.
Catalog number 041093.KS
Price (EUR)
1.245,00
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (EUR)
1.245,00
Each
Specifications
Assay Percent Notes(metals basis)
CAS10097-28-6
ChEBICHEBI:30588
Chemical Name or MaterialSilicon(II) oxide sputtering target
EINECS Number233-232-8
View more
Silicon(II) oxide sputtering target coatings are used for optical applications in reflectors, flood lamps, mirrors, jewelry, and other products. The coatings can reduce reflection in the near infrared range or act as interference layers. It is used in thin-film capacitors, hybrid circuits, semiconductor components and other electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only