Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99.9% (metals basis)
Thermo Scientific Chemicals

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99.9% (metals basis)

CAS: 14040-05-2 | C22H38CuO4 | 430.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 041279.03
also known as 041279-03
Price (EUR)
81,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
81,10
Each
Chemical Identifiers
CAS14040-05-2
IUPAC Namecopper(2+) bis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Molecular FormulaC22H38CuO4
InChI KeyJGMGCTZGOAONPQ-UHFFFAOYSA-N
SMILES[Cu++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark violet or purple to dark purple or dark green
FormCrystals or powder or chunks
Complexometric Titration%Cu (14.65-14.75%)
Total Metal Impurities≤0.1% (≤1000ppm)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only