Xylenol Orange tetrasodium salt, ACS
Xylenol Orange tetrasodium salt, ACS
Xylenol Orange tetrasodium salt, ACS
Thermo Scientific Chemicals

Xylenol Orange tetrasodium salt, ACS

CAS: 3618-43-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 041379.09
also known as 041379-09
Price (EUR)
243,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
243,00
Each
Chemical Identifiers
CAS3618-43-7
IUPAC Nametetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular FormulaC31H28N2Na4O13S
InChI KeyFXYMNKLSOQONJB-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(C2=CC=CC=C2OS1(=O)=O)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentClarity of solution P.T.
CommentSuitability for zinc titration P.T.
Xylenol Orange tetrasodium salt is used as a photometric reagent and a metal indicator. It is a versatile indicator for EDTA titrations in acidic range. It is a common organic reagent used as an indicator for metal titrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only