Silver wire, 1.0mm (0.04in) dia, hard, 99.9% (metals basis)
Silver wire, 1.0mm (0.04in) dia, hard, 99.9% (metals basis)
Silver wire, 1.0mm (0.04in) dia, hard, 99.9% (metals basis)
Silver wire, 1.0mm (0.04in) dia, hard, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver wire, 1.0mm (0.04in) dia, hard, 99.9% (metals basis)

Form: Approx.8.24g/m; Temper: hard | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
25 m
Catalog number 041456.G1
also known as 041456-G1
Price (EUR)
-
Quantity:
1 m
Request bulk or custom format
Specifications
AppearanceSilver-colored wire
Assay Percent Range99.9% (Metals basis)
CAS7440-22-4
ChEBICHEBI:9141
Chemical Name or MaterialSilver
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only