1-Methoxy-2-propanol, 99+%
1-Methoxy-2-propanol, 99+%
1-Methoxy-2-propanol, 99+%
Thermo Scientific Chemicals

1-Methoxy-2-propanol, 99+%

CAS: 107-98-2 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 L
4 x 4 L
Catalog number 041547.M6
also known as 041547-M6
Price (EUR)
400,00
Each
Quantity:
4 x 4 L
Request bulk or custom format
Price (EUR)
400,00
Each
Chemical Identifiers
CAS107-98-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Assay (GC)> 99.0%
Comment2-Methoxy-1-propanol content: 0.49% max
Water Content (Karl Fischer Titration)0.1%, max
Formliquid
1-Methoxy-2-propanol is used as a reagent in the synthesis of 2-amino-3-carboxy-4-phenylthiophenes, which acts as a protein kinase C inhibitors. It is also used as a reagent in the synthesis of metolachlor. It acts as a good biological indicator. Further, it is used as a solvent and an antifreeze agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only