Zinc sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)
Zinc sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)
Zinc sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Zinc sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
041656.KS1 Ea.
Catalog number 041656.KS
Price (EUR)
476,00
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (EUR)
476,00
Each
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSputtering target, 50.8mm (2.0in) diameter x 6.35mm (0.250in) thick.
Appearance (Color)Gray
Assay from Supplier's CofA≥99.99% (metals basis)
Zinc sputtering target is used in the manufacture of paints, bleaches and to make other chemicals. It is also used in medical research. Pure zinc is used as an electrode in a Daniel cell and also in dry batteries. It is used as a galvanic coating on steel to prevent corrosion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only