Antimony, plasma standard solution, Specpure™ Sb 10,000μg/mL
Antimony, plasma standard solution, Specpure™ Sb 10,000μg/mL
Antimony, plasma standard solution, Specpure™ Sb 10,000μg/mL
Thermo Scientific Chemicals

Antimony, plasma standard solution, Specpure™ Sb 10,000μg/mL

Sb in H2O/tartaric acid/tr. HNO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 041683.AD
also known as 041683-AD
Price (EUR)
80,80
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
80,80
Each
Specifications
Chemical Name or MaterialAntimony, plasma standard solution, Specpure™
MDL NumberMFCD00134030
Molecular FormulaSb in H2O/tartaric acid/tr. HNO3
Physical FormLiquid
Quantity50 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only