Calcium boride, 99.5% (metals basis)
Calcium boride, 99.5% (metals basis)
Calcium boride, 99.5% (metals basis)
Thermo Scientific Chemicals

Calcium boride, 99.5% (metals basis)

CAS: 12007-99-7 | B6Ca | 104.938 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 041722.06
also known as 041722-06
Price (EUR)
69,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
69,50
Each
Chemical Identifiers
CAS12007-99-7
IUPAC Namecalcium pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide
Molecular FormulaB6Ca
InChI KeyAWVIDOFQNLOVRC-UHFFFAOYSA-N
SMILES[Ca++].B12B3[B-]11B4B1[B-]234
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, -325 mesh
Total Metal Impurities≤0.5%
Calcium boride is used as an electrode (cathode) material and n-type thermoelectric materials. It is used to make boron- alloyed steel, as a deoxidation agent in the production of oxygen-free copper. It is further used as an antioxidant in carbon bonded refractories. It acts as a new material which is used in the nuclear-industry for neutron- preventing, high-purity metal borid (TiB2,ZrB2,HfB2) and high-purity boron alloy(Ni-B, Co-B, Cu-b).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only