Chloroacetic acid, ACS, 99%
Chloroacetic acid, ACS, 99%
Chloroacetic acid, ACS, 99%
Thermo Scientific Chemicals

Chloroacetic acid, ACS, 99%

CAS: 79-11-8 | C2H3ClO2 | 94.494 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
4 x 500 g
Catalog number 041724.36
also known as 041724-36
Price (EUR)
97,20
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
97,20
Each
Chemical Identifiers
CAS79-11-8
IUPAC Name2-chloroacetic acid
Molecular FormulaC2H3ClO2
InChI KeyFOCAUTSVDIKZOP-UHFFFAOYSA-N
SMILESOC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCl 0.01%
Impurity contentSO₄ 0.02%
Trace Metals(as Pb) 0.001%
Residue on ignition0.02%
Impurity contentFe 0.002%
View more
Chloroacetic acid is used in the production of pharmaceuticals, dyes, detergents, synthetic caffeine, glycine and thioglycolic acid, which acts as a stabilizer in PVC. It acts as a precursor to glyphosate and adrenaline. It also serves as a preservative and bacteriostat. Further, it is used as surface active agents and photosensitive chemicals. It is also used in the O-alkylation of salicylaldehyde and an active component in cosmetics. In addition, it is involved in the preparation of thickening agent carboxymethyl cellulose and carboxymethyl starch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only