Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles
Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles
Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles

CAS: 209733-38-0 | C15H35MoO5 | 391.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 041836.AE
also known as 041836-AE
Price (EUR)
488,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
488,00
Each
Specifications
Assay Percent Notes(metals basis)
CAS209733-38-0
Chemical Name or MaterialMolybdenum(V) isopropoxide
ColorYellow to Brown
Concentration or Composition (by Analyte or Components)5% w/v in isopropanol
View more
Used in the preparations of nanomaterials and thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only