Cerium(III) sulfide, 99.9% (metals basis)
Cerium(III) sulfide, 99.9% (metals basis)
Cerium(III) sulfide, 99.9% (metals basis)
Thermo Scientific Chemicals

Cerium(III) sulfide, 99.9% (metals basis)

CAS: 12014-93-6 | Ce2S3 | 376.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 041856.06
also known as 041856-06
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS12014-93-6
IUPAC Namedicerium(3+) trisulfanediide
Molecular FormulaCe2S3
InChI KeyMMXSKTNPRXHINM-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[Ce+3].[Ce+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Used as pharmaceutical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only