Thionyl chloride, 99%, Thermo Scientific Chemicals
Thionyl chloride, 99%, Thermo Scientific Chemicals
Thionyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thionyl chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2 x 1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041868.36
Price (EUR)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7719-09-7
IUPAC Namesulfurooyl dichloride
Molecular FormulaCl2OS
InChI KeyFYSNRJHAOHDILO-UHFFFAOYSA-N
SMILESClS(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥98.5%
Thionyl chloride is used as a chlorinating reagent for acids and alcohols. It is often used in the preparation of a wide variety of alkyl chloride and acyl chloride compounds, which are useful in the manufacture of fine chemicals, drugs, drug intermediates and vitamins. It is used to prepare thionyl bromide and thionyl fluoride. It finds use in several important organic transformations including pummerer rearrangement, and in lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only