Lithium bis(trimethylsilyl)amide, 0.9-1.1M in hexane, packaged under Argon in resealable ChemSeal™ bottles
Lithium bis(trimethylsilyl)amide, 0.9-1.1M in hexane, packaged under Argon in resealable ChemSeal™ bottles
Lithium bis(trimethylsilyl)amide, 0.9-1.1M in hexane, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 0.9-1.1M in hexane, packaged under Argon in resealable ChemSeal™ bottles

CAS: 4039-32-1 | C6H18LiNSi2 | 167.327 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 041872.22
also known as 041872-22
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS4039-32-1
IUPAC Namelithium(1+) bis(trimethylsilyl)azanide
Molecular FormulaC6H18LiNSi2
InChI KeyYNESATAKKCNGOF-UHFFFAOYSA-N
SMILES[Li+].C[Si](C)(C)[N-][Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid
Concentration0.90-1.10M
Lithium bis(trimetylsilyl)amide is used as a strong base in organic synthesis and as catalysts in polymerization reactions. It is also used to form various organolithium compounds such as acetylides and lithium enolates. It is a base employed in generating enolates for preparation of lactone precursors, pyranones and cyclohexanes. Further, it is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. In addition, it is used in a three-step synthesis of disubstituted 1,2,5-thiadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only