Sulfur in Crude Oil standard solution, Specpure™, 10,000μg/g (1.00%)
Thermo Scientific Chemicals

Sulfur in Crude Oil standard solution, Specpure™, 10,000μg/g (1.00%)

 
Have Questions?
Catalog NumberQuantity
041888.AE
also known as 041888-AE
100 mL
Catalog number 041888.AE
also known as 041888-AE
Price (EUR)
254,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
254,00
Each
Specifications
Chemical Name or MaterialSulfur in Crude Oil standard solution
ColorBlack
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM CRUDE OIL
Health Hazard 1H226-H350
Health Hazard 2GHS H Statement
H350-H226
May cause cancer.
Flammable liquid and vapor.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only