Polystyrene standard, M.W. 650,000, Mw/Mn 1.06
Polystyrene standard, M.W. 650,000, Mw/Mn 1.06
Polystyrene standard, M.W. 650,000, Mw/Mn 1.06
Thermo Scientific Chemicals

Polystyrene standard, M.W. 650,000, Mw/Mn 1.06

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
041941.03
also known as 041941-03
1 g
Catalog number 041941.03
also known as 041941-03
Price (EUR)
89,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
89,30
Each
Specifications
CAS9003-53-6
ChEBICHEBI:27452
Chemical Name or MaterialPolystyrene standard
IUPAC Namestyrene
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only