Zinc selenide, 99.99% (metals basis)
Zinc selenide, 99.99% (metals basis)
Zinc selenide, 99.99% (metals basis)
Thermo Scientific Chemicals

Zinc selenide, 99.99% (metals basis)

CAS: 1315-09-9 | SeZn | 144.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 041958.14
also known as 041958-14
Price (EUR)
186,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
186,00
Each
Chemical Identifiers
CAS1315-09-9
IUPAC Namezinc(2+) selandiide
Molecular FormulaSeZn
InChI KeyGTLQJUQHDTWYJC-UHFFFAOYSA-N
SMILES[Zn++].[Se--]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only