Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.99% (metals basis), Thermo Scientific Chemicals
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.99% (metals basis), Thermo Scientific Chemicals
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.99% (metals basis), Thermo Scientific Chemicals

MOCVD of zirconium
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042028.14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS18865-74-2
Chemical Name or MaterialTetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV)
Formula Weight824.3
Health Hazard 1H315-H319-H335
View more
MOCVD of zirconium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only